Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIBCH Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514336

Catalog No. PIPA514336

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

HIBCH is responsible for the specific hydrolysis of HIBYL-CoA, a valine catabolite, as well as the hydrolysis of beta-hydroxypropionyl-CoA, an intermediate in a minor pathway of propionate metabolism.
Specifications

Specifications

HIBCH
Polyclonal
PBS with 0.09% sodium azide
Q6NVY1
3-hydroxyisobutyryl-coenzyme A hydrolase; HIB-CoA hydrolase; HIBYL-CoA-H; 3-hydroxyisobutyryl-CoA hydrolase, mitochondrial; HIBCH
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26275
Immunohistochemistry, Western Blot
Unconjugated
HIBCH
Liquid
HIBCH
KLH conjugated synthetic peptide between 183-209 amino acids from the central region of human HIBCH
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit