Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-1 alpha Rabbit anti-Human, DyLight 550, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP147180R

Catalog No. NBP147180R


Add to cart

Description

Description

HIF-1 alpha Polyclonal antibody specifically detects HIF-1 alpha in Human samples. It is validated for Western Blot.
Specifications

Specifications

HIF-1 alpha
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
HIF1A
This HIF-1 alpha Antibody - Exon 12 was developed against a synthetic peptide made to an internal portion of the human HIF-1 alpha protein (within amino acids 550 - 600) [Uniprot# Q16665].
93 kDa
0.1 ml
Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators
Primary
3091
Human
Western Blot
Western Blot
Q16665
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
HIF-1 alpha Antibody - Exon 12 is specific for Exon 12 of HIF-1 alpha.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only