Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HIF-3 alpha, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $314.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Bacterially expressed human HIF-3 alpha (N-terminus). [UniProt# Q9Y2N7]
Antigen HIF-3 alpha
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1002287SS
View Documents
Novus Biologicals
NB1002287SS
0.025 ml
Each for $119.60
Add to cart
 
NB1002287
View Documents
Novus Biologicals
NB1002287
0.1 ml
Each for $314.00
Add to cart
 
Description

Description

HIF-3 alpha Polyclonal antibody specifically detects HIF-3 alpha in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Rabbit
Bacterially expressed human HIF-3 alpha (N-terminus). [UniProt# Q9Y2N7]
Polyclonal
Primary
Western Blot
64344
1 mg/mL
IgG
HIF-3 alpha
Affinity Purified
RUO
Q9Y2N7
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit