Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF1AN Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511150

Catalog No. PIPA511150

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with zebrafish based on sequence homology.

FIH1Encoded protein (factor inhibiting HIF-1)is a co-repressor that interacts with hypoxia-inducible factor 1(HIF-1) alpha and the von Hippel-Lindau tumor suppressor protein to mediate repression of HIF-1 transcriptional activity.
Specifications

Specifications

HIF1AN
Polyclonal
PBS with 0.09% sodium azide
Q9NWT6, Q8BLR9
Hypoxia-inducible factor 1 alpha inhibitor; Hypoxia-inducible factor asparagine hydroxylase; Factor inhibiting HIF-1; FIH-1; HIF1AN; FIH1;
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
319594, 55662
Western Blot
Unconjugated
HIF1AN
Liquid
HIF1AN
KLH conjugated synthetic peptide between 286-316 amino acids from the C-terminal region of human HIF1AN
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit