Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HIN-1/SCGB3A1, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136983

Catalog No. NBP136983


Add to cart

Description

Description

HIN-1/SCGB3A1 Polyclonal antibody specifically detects HIN-1/SCGB3A1 in Human samples. It is validated for Peptide ELISA.
Specifications

Specifications

HIN-1/SCGB3A1
0.5 mg/mL
Peptide ELISA Detection limit 1:32000
NP_443095.2
cytokine high in normal-1, Cytokine HIN-1, High in normal 1, HIN-1, HIN1Pneumo secretory protein 2, LU105, PNSP2, PnSP-2Uteroglobin-related protein 2, secretoglobin family 3A member 1, secretoglobin, family 3A, member 1, UGRP2MGC87867
Goat
IgG
0.1 mg
Cell Cycle and Replication
Primary
92304
Human
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
SCGB3A1
Peptide with sequence NHLIEGSQKCVAE corresponding to internal region according to NP_443095.2.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody recognizes and epitope within the internal.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only