Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HINFP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB10056628SS

 View more versions of this product

Catalog No. NB10056628T


Add to cart

Description

Description

HINFP Polyclonal antibody specifically detects HINFP in Human samples. It is validated for Western Blot.
Specifications

Specifications

HINFP
0.5 mg/mL
Western Blot 1:500
Q9BQA5
HiNF-PDKFZP434F162, histone H4 gene-specific protein HiNF-P, histone H4 transcription factor, Histone nuclear factor P, MBD2 (methyl-CpG-binding protein)-interacting zinc finger protein, MBD2-interacting zinc finger 1, MBD2-interacting zinc finger protein, Methyl-CpG-binding protein 2-interacting zinc finger protein, MIZFDKFZp434F162, ZNF743MBD2-interacting zinc finger
Rabbit
IgG
Protein G purified
RUO
Primary
25988
Human
Western Blot
Unconjugated
PBS containing 0.2% gelatin with 0.02% Sodium Azide
Antisera
HINFP
This antibody was developed against a mixture of synthetic peptides corresponding to amino acid residues, 180-194, 331-346, and 371-388 of human MIZF.
64 kDa
0.05 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
In MOLT-4, a 64 kDa band is observed.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only