Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HINT1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human HINT1 protein.
Supplier: Abnova Corporation H00003094B01P
Description
Sequence: MADEIAKAQVARPGGDTIFGKIIRKEIPAKIIFEDDRCLAFHDISPQAPTHFLVIPKKHISQISVAEDDDESLLGHLMIVGKKCAADLGLNKGYRMVVNEGSDGGQSVYHVHLHVLGGRQMHWPPGSpecifications
HINT1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human HINT1 protein. | |
HINT1 | |
FLJ30414/FLJ32340/HINT/PKCI-1/PRKCNH1 | |
Mouse | |
Affinity chromatography | |
RUO | |
3094 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005340.3 | |
HINT1 | |
HINT1 (NP_005331.1, 1 a.a. ∼ 126 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction