Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hip1R Mouse, Unlabeled, Clone: 44, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612118

Catalog No. BDB612118


Add to cart

Description

Description

Clathrin is the major protein component in the coat formed around pits and vesicles involved in receptor-mediated endocytosis. Clathrin forms a non-covalently bound triskelion structure composed of three heavy chains (192kDa each) and three light chains (23-25kDa each). A variety of proteins facilitate receptor-mediated endocytosis through association with clathrin-coated vesicles. Huntingtin interacting protein 1 (Hip1) is an actin-binding protein that interacts with Huntingtin protein, and has been implicated in vesicular transport defects found in Huntingtin's disease. Hip1 related protein (Hip1R) is another actin binding protein that contains an epsin NH2-terminal homology (ENTH)domain, three coiled-coil regions, a leucine zipper, and a talin-like actin binding domain. Hip1R mRNA is widely expressed, and Hip1R protein is enriched in the cell cortex and perinuclear region. The ENTH domain of Hip1R is required for binding to phosphatidylinositol-4,5-bisphosphate, and this complex is essential for clathrin-mediated endocytosis. In addition, Hip1R colocalizes with clathrin, AP-2 and endocytosed transferrin. Thus, Hip1R may facilitate interactions between clathrin-coated pits and actin during endocytosis.

Western Blotting

Specifications

Specifications

Hip1R
44
Unconjugated
Affinity Purified
Mouse Hip1R aa. 560-772
50μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit