Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
HIP2 Mouse anti-Human, Clone: 1D10, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519073
Catalog No. PIMA519073
Description
Description
Peptide Sequence: MLAMSPPAAD SYPPPPPPLR PPPPPPPLPP PPPPSAIQT*
The protein encoded by this gene belongs to the ubiquitin-conjugating enzyme family. This protein interacts with RING finger proteins, and it can ubiquitinate huntingtin, the gene product for Huntington ft.s disease. Known functions for this protein include a role in aggregate formation of expanded polyglutamine proteins and the suppression of apoptosis in polyglutamine diseases, a role in the dislocation of newly synthesized MHC class I heavy chains from the endoplasmic reticulum, and involvement in foam cell formation. Multiple transcript variants encoding different isoforms have been identified for this gene.
Specifications
Specifications
HIP2
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P61086
UBE2K; HIP2; LIG; Ubiquitin-conjugating enzyme E2 K; Huntingtin-interacting protein 2; Ubiquitin carrier protein; Ubiquitin-conjugating enzyme E2-25 kDa; Ubiquitin-protein ligase
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3093
ELISA, Western Blot
1D10
Unconjugated
UBE2K
Liquid
UBE2K
UBE2K (-, 1 a.a. ∽ 40 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human