Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIP55 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528400

Catalog No. PIPA528400


Add to cart

Description

Description

PA5-28400 targets HIP55 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-28400 immunogen is synthetic peptide corresponding to a region within amino acids 1 and 44 of Human HIP55. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Actin-binding adapter protein. May act as a common effector of antigen receptor-signaling pathways in leukocytes. Its association with dynamin suggests that it may also connect the actin cytoskeleton to endocytic function. Acts as a key component of the immunological synapse that regulates T-cell activation by bridging TCRs and the actin cytoskeleton to gene activation and endocytic processes. Binds to F-actin but is not involved in actin polymerization, capping or bundling. Does not bind G-actin.
Specifications

Specifications

HIP55
Polyclonal
Unconjugated
DBNL
Liquid
DBNL
Synthetic peptide corresponding to a region within amino acids 1 and 44 of Human HIP55
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 10% glycerol and 0.01% thimerosal; pH 7
Q9UJU6
ABP1, HIP-55, SH3P7, HPK1-interacting protein of 55 kDa, SH3 domain-containing protein 7, cervical SH3P7, cervical mucin-associated protein, drebrin-F, drebrin-like protein, src homology 3 domain-containing protein HIP-55
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
28988
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit