Learn More
anti-HIPK1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP133658
Catalog No. NBP133658
Add to cart
Description
Description
HIPK1 Polyclonal specifically detects HIPK1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
HIPK1
0.25mg/mL
Western Blot 1:500-1:3000
Affinity Purified
HIPK1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human HIPK1. The exact sequence is proprietary.
131 kDa
0.1 ml
Protein Kinase
Primary
204851
Human, Mouse
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
EC 2.7.11.1, homeodomain interacting protein kinase 1, homeodomain interacting protein kinase 1-like protein, homeodomain-interacting protein kinase 1, KIAA0630MGC26642, MGC33446, MGC33548, Myak, Nbak2, nuclear body associated kinase 2b
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Cross reactivity in Mouse determined by Western Blot. Not yet tested in any other species.