Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HISPPD1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232155

Catalog No. NBP232155


Add to cart

Description

Description

HISPPD1 Polyclonal antibody specifically detects HISPPD1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

HISPPD1
1.0 mg/mL
Western Blot 1:1000 - 1:5000, Immunoprecipitation 2 - 10 ug/mg lysate
BAA24863.2
diphosphoinositol pentakisphosphate kinase 2FLJ21506, EC 2.7.4.21, EC 2.7.4.24, HISPPD1histidine acid phosphatase domain containing 1, Histidine acid phosphatase domain-containing protein 1, hsVIP2, inositol heptaphosphate kinase 2, inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2, InsP6 and PP-IP5 kinase 2, KIAA0433FLJ23463, VIP1 homolog 2, VIP2IP7K2
Rabbit
IgG
100 ul
Protein Phosphatase
Primary
23262
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
PPIP5K2
The immunogen this antibody was made to, maps to a region between residue 925 to 975 of human Diphosphoinositol Pentakisphosphate Kinase 2 using the numbering given in entry BAA24863.2 (GeneID 23262).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only