Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H1.2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526171

Catalog No. PIPA526171

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Histones H1 are necessary for the condensation of nucleosome chains into higher order structures.
Specifications

Specifications

Histone H1.2
Polyclonal
PBS with 0.09% sodium azide
P16403
H1.2, H1C, H1F2, H1 histone family, member 2, histone 1, H1c, histone H1.2, histone H1d
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3006
Western Blot
Unconjugated
HIST1H1C
Liquid
HIST1H1C
KLH conjugated synthetic peptide between 105-131 amino acids from the central region of human HIST1H1C
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit