Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST1H2BJ Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524220

Catalog No. PIPA524220


Add to cart

Description

Description

This antibody is predicted to react with bovine, chicken, mouse, non-human primate, rat and Xenopus and zebrafish based on sequence homology.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A,H2B, H3, and H4) form an octamer, around which approximately 146 bpof DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H2B family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the histone microcluster on chromosome 6p21.33.
Specifications

Specifications

HIST1H2BJ
Polyclonal
PBS with 0.09% sodium azide
P06899
H2B/r, H2BFR, H2B histone family, member R, histone 1, H2bj, histone H2B type 1-J, histone H2B.1, histone H2B.r
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8970
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HIST1H2BJ
Liquid
HIST1H2BJ
KLH conjugated synthetic peptide between 57-86 amino acids from the central region of human HIST1H2BJ
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit