Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HRH3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521312
Description
Recommended positive controls: HeLa, Raji. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HRH3 Polyclonal antibody specifically detects HRH3 in Human samples. It is validated for Western Blot.Specifications
HRH3 | |
Polyclonal | |
Unconjugated | |
HRH3 | |
AW049250; G protein-coupled receptor; G protein-coupled receptor 97; GPCR97; G-protein coupled receptor 97; H3R; HH3R; histamine H3 receptor; histamine H3 receptor H3L isoform; histamine H3 receptor H3S isoform; histamine receptor; histamine receptor H 3; histamine receptor H3; Hrh3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11255 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y5N1 | |
HRH3 | |
Synthetic peptide corresponding to a region within amino acids 299 and 389 of Histamine H3 Receptor (Uniprot ID#Q9Y5N1) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction