Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRH4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA124553

Catalog No. PIPA124553

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

PA1-24553 detects Histamine H4 Receptor in human samples. PA1-24553 has been successfully used in Immunocytochemistry procedures. The PA1-24553 immunogen corresponds to a synthetic peptide, KLH conjugated, corresponding to the C-terminal domain of human Histamine H4 Receptor. Sequence identity/similarity to Mouse: 58/76%, Rat: 35/41%.

Histamine Receptor H4 binds histamine and several psychoactive compounds such as amitriptyline, chlorpromazine, cyproheptadine, and mianserin. The Histamine H4 receptor has been reported in human blood peripheral leukocytes, bone marrow, colon, liver, lung, smalll intestine, spleen, testis, thymus, tonsil, and trachea. Contrary to the mouse H4 receptor, the human gene was not detected in brain.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HRH4
Polyclonal
Unconjugated
Q9H3N8
HRH4
Synthetic peptide, KLH conjugated, corresponding to the C-terminal domain of human Histamine H4 Receptor.
25 μg
Primary
Human
Antibody
IgG
Immunocytochemistry, Immunofluorescence
1 mg/mL
HRH4
AXOR35, BG26, GPCR105, GPRv53, H4, H4R, HH4R, MGC130500
Rabbit
Antigen affinity chromatography
RUO
59340
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit