Learn More
Histamine Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136089
Catalog No. 01-672-266
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide. For long-term storage, aliquot and store at -20°C.
The biogenic amine histamine is an important modulator of numerous physiologic processes, including neurotransmission, gastric acid secretion, and smooth muscle tone.
Specifications
Specifications
Histamine
Polyclonal
whole serum with no preservative
Rabbit
IgG
RUO
Antibody
Polyclonal
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
Lyophilized
Histamine conjugated to HSA.
50 μL
4° C
Primary
Rat