Learn More
HDC Rabbit anti-Canine, Guinea Pig, Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136090
Catalog No. 01-672-267
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide. For long-term storage, aliquot and store at -20°C.
The biogenic amine histamine is an important modulator of numerous physiologic processes, including neurotransmission, gastric acid secretion, and smooth muscle tone. The biosynthesis of histamine from histidine is catalyzed by the enzyme L-histidine decarboxylase (HDC; EC 4.1.1.22). This homodimeric enzyme is a pyridoxal phosphate (PLP)-dependent decarboxylase and is highly specific for its histidine substrate.[supplied by OMIM]
Specifications
Specifications
HDC
Polyclonal
whole serum with no preservative
P19113, P23738, P16453
MGC163399
Rabbit
IgG
RUO
Antibody
Polyclonal
Canine, Guinea Pig, Human, Mouse, Rat
Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
HDC
Lyophilized
HDC
Recombit histidine decarboxylase produced in E. coli. .
50 μL
4° C
Primary
100719978, 15186, 24443, 3067, 487551