Learn More
Histidine Tagged Mouse anti-All, Clone: 4D11, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05531
Catalog No. 05-531-MI
Add to cart
Description
Description
This Anti-Histidine Tagged Antibody (anti His antibody) clone 4D11 is validated for use in immunocytochemistry, western blotting for the detection of Histidine Tagged.
Specifications
Specifications
Histidine Tagged
4D11
Protein G Purified immunoglobulin in Immunoaffinity Purified immunoglobulin in 0.014M phosphate buffer, pH 7.6, 0.175M NaCl, and 0.07% sodium azide.
Murine
κ
200μg
Primary
All
Immunocytochemistry, Western Blot
Unconjugated
Purified
N-terminal His-tagged and truncated TRAP protein corresponding to amino acids 26-248 from P. falciparum expressed in E coli.
Protein G purfied
2-8°C for one year
Monoclonal