Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141614

Catalog No. 01-675-230


Add to cart

Description

Description

Suggested positive control: PBMC.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromotin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transciption regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.
Specifications

Specifications

Histone H2B
Polyclonal
Unconjugated
HIST1H2BB
Liquid
HIST1H2BB
Synthetic peptide corresponding to a portion of human Histone-H2B.
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P33778
RP1-193B12.10, H2B/d, H2BFD, HIST1H3I, MGC125414, MGC125415, MGC125416, MGC9388
Rabbit
IgG
200 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3018
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit