Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3.3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527507

Catalog No. PIPA527507


Add to cart

Description

Description

PA5-27507 targets Histone H3.3 in IF and WB applications and shows reactivity with Human and mouse samples. The PA5-27507 immunogen is recombit protein fragment corresponding to a region within amino acids 1 and 105 of Human Histone H3.3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene contains introns and its mRNA is polyadenylated, unlike most histone genes. The protein encoded is a replication-independent member of the histone H3 family.
Specifications

Specifications

Histone H3.3
Polyclonal
Unconjugated
H3F3A
Liquid
H3F3A
Recombit protein fragment corresponding to a region within amino acids 1 and 105 of Human Histone H3.3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
ChIP assay, Immunocytochemistry, Immunofluorescence, Western Blot
0.08 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P84243, P84244
Histone H3, H3.3A, H3.3B, H3F3, H3F3A, H3F3B
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
15078, 3020
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit