Learn More
Histone H3 Rabbit anti-Human, Chicken, Rat, Mouse, Yeast (S. cerevisiae), Clone: A3S, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma 05928
Catalog No. 05-928-MI
Add to cart
Description
Description
Use Anti-Histone H3 Antibody, CT, pan, clone A3S (Rabbit Monoclonal Antibody) validated in Chimmunoprecipitation, western blotting to detect Histone H3 also known as Histone H3.
Specifications
Specifications
Histone H3
A3S
Purified Rabbit Monoclonal IgG in buffer containing 0.014M phosphate buffer, pH 7.6, 0.175M NaCl, 0.07% sodium azide and 30% glycerol.
Purified
KLH-conjugated, synthetic peptide corresponding to the C-terminus of human Histone H3.
Protein A purfied
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Monoclonal
Chicken, Human, Murine, Rat, S. cerevisiae, Yeast
Chromatin Immunoprecipitation, Western Blot
Unconjugated
NM_003493
Rabbit
IgG
100μL
Primary
H3F3A, MGC87783, H3.3A, MGC87782, H3F3, H3.3B, H3F3B