anti-Histone H3, Clone: CMA301, EMD Millipore™

Manufacturer: EMD Millipore 051341S

 View more versions of this product

Catalog No. 05-134-1SMI

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Immunogen Synthetic peptide corresponding to amino acids 1-12 of human Histone H3, conjugated to KLH.
    Isotype κ
    Formulation Mouse monoclonal IgG2bκ supernatant in PBS with 0.05% sodium azide.
    Applications Dot Blot,ELISA,Flow Cytometry,Immunocytochemistry,Immunoprecipitation,Western Blotting
    Host Species Mouse
    Storage Requirements 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
    Clone CMA301
    Gene Accession No. NM_003493
    Purification Method Peptide affinity chromatography
    Primary or Secondary Primary
    Conjugate Unlabeled
    Antigen Histone H3
    Monoclonal or Polyclonal Monoclonal
    Cross Reactivity Human,Vertebrate
    Format Affinity Purified
    Gene Symbols H3.4, H3/g , H3/t, H3FT, H3T, H3t, MGC126886, MGC126888, OTTHUMP00000037945
    Quantity 10μg

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit