Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Histone H3, CT, pan, Clone: A3S, ™ (Upstate™)
Supplier: MilliporeSigma™ 05928/DEL

Description
An antibody that recognizes the C-terminus of histone H3 and is useful for identifying total H3 independent of post-translational modifications. GenBank Number: H3F3B. MW: 17kDa.
Chromatin Immunoprecipitation, Western Blotting
Specifications
Histone H3, CT, pan | |
Unconjugated | |
KLH-conjugated, synthetic peptide corresponding to the C-terminus of human Histone H3 | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Rabbit | |
100 μL | |
Epigenetics | |
Chicken, Human, Mouse, Rat, Yeast | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title