Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, p Thr3, Monomethyl Lys4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB211012
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Dot Blot, Chromatin Immunoprecipitation (Negative).Specifications
Histone H3 (p Thr3, Monomethyl Lys4) | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K4Me1, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
Rabbit | |
15 kDa | |
0.05 mg | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
126961 | |
Human, Mouse, C. elegans | |
IgG |
ChIP Assay, Dot Blot, Immunocytochemistry, Immunofluorescence | |
0.98 mg/mL | |
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:100, Dot Blot 1:1000, Chromatin Immunoprecipitation (Negative) | |
Q71DI3 | |
H3C14 | |
This Histone H3 [p Thr3, Monomethyl Lys4] antibody was raised against synthetic monomethylated/phosphorylated peptides surrounding Lysine 4 and Threonine 3 of human Histone H3.2 [Swiss Prot Q71DI3]. | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction