Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, Trimethyl Lys18 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
$265.00 - $409.50
Specifications
Antigen | Histone H3 (Trimethyl Lys18) |
---|---|
Concentration | 0.6 mg/mL |
Dilution | Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:50, Dot Blot 0.5 ug/ml, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Dot Blot, ChIP Assay |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB211143SS
|
Novus Biologicals
NB211143SS |
0.025 mg |
Each for $265.00
|
|
|||||
NB211143
|
Novus Biologicals
NB211143 |
0.05 mg |
Each for $409.50
|
|
|||||
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Dot Blot.Specifications
Histone H3 (Trimethyl Lys18) | |
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:50, Dot Blot 0.5 ug/ml, Chromatin Immunoprecipitation (ChIP) 2-5 ug per million cells | |
Polyclonal | |
Rabbit | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K18Me3, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
H3C14 | |
IgG | |
Affinity Purified | |
Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology. |
0.6 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Dot Blot, ChIP Assay | |
Unconjugated | |
RUO | |
Human, Mouse, C. elegans | |
Q71DI3, P84228 | |
126961 | |
This Histone H3 [Trimethyl Lys18] antibody was raised against synthetic trimethylated peptide surrounding Lysine 18 of human Histone H3.2 [Swiss Prot Q71DI3]. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
15 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title