Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HIV-I gp41, Clone: 10E9, ZeptoMetrix

Murine monoclonal antibody reacts with Human Immunodeficiency Virus Type 1 (HIV-1) gp41 envelope transmembrane protein

Supplier:  ZeptoMetrix 0801006

 View more versions of this product

Catalog No. 22-157-317


Add to Cart

Description

Description

Host Species: Mouse
Clone: 10E9;
Isotype: IgG1
Species Reactivity: Hu

Enzyme Immunoassay, Immunofluorescence, Western Blotting

Specifications

Specifications

HIV-I gp41
10E9
Mouse
RUO
Human
IgG1
Monoclonal
Unconjugated
100 μg
Primary
Purified
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit