Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HIV-I p17, Clone: 2D11, ZeptoMetrix™ Corporation

Murine monoclonal antibody reacts with Human Immunodeficiency Virus Type 1 (HIV-1) p17 protein

$286.00 - $1822.00

Specifications

Antigen HIV-I p17
Clone 2D11
Format Purified
Host Species Murine
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
22-157-329
View Documents
ZeptoMetrix Corporation
0801076
100μg
Each for $286.00
Add to cart
 
22-157-352
View Documents
ZeptoMetrix Corporation
0801122
1mg
Each for $1,822.00
Add to cart
 
Description

Description

Host Species: Mouse
Clone: 2D11
Isotype: IgG1
Species Reactivity: Hu

Enzyme Immunoassay, Immunofluorescence, Western Blotting

Specifications

Specifications

HIV-I p17
Purified
RUO
Monoclonal
2D11
Murine
IgG1
Primary
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit