Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HIV-I p17, Clone: 32/1.24.89, ZeptoMetrix™ Corporation

Murine monoclonal antibody reacts with Human Immunodeficiency Virus Type 1 (HIV - 1) p17 core protein

Manufacturer:  ZeptoMetrix Corporation 0801115

 View more versions of this product

Catalog No. 22-156-772


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Host Species: Mouse
Clone: 32/1.24.89
Isotype: IgG1
Species Reactivity: Hu

Enzyme Immunoassay, Immunofluorescence, Western Blotting

Specifications

Specifications

HIV-I p17
Unconjugated
Murine
1mg
Primary
Human
32/1.24.89
Purified
IgG1
RUO
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit