anti-HIV-I p24, Clone: 39/5.1.23, ZeptoMetrix™ Corporation Available on GSA/VA Contract for Federal Government customers only.

Murine monoclonal antibody reacts with Human Immunodeficiency Virus Type 1 (HIV-1) p24 protein

Manufacturer: ZeptoMetrix CorporationSupplier Diversity Partner 0801126

 View more versions of this product

Catalog No. 22-157-356

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Antigen HIV-I p24
    Format Purified
    Host Species Mouse
    Isotype IgG1
    Quantity 1mg
    Regulatory Status RUO
    Primary or Secondary Primary
    Monoclonal or Polyclonal Monoclonal
    Clone 39/5.1.23
    Conjugate Unlabeled
    Cross Reactivity Human

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit