Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HIV-I p24, Clone: 39/5.4A, ZeptoMetrix

Murine monoclonal antibody reacts with Human Immunodeficiency Virus Type 1(HIV-1) p24 protein

$296.50 - $2030.00

Specifications

Antigen HIV-I p24
Clone 39/5.4A
Format Purified
Host Species Mouse
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
22-157-332
View Documents
ZeptoMetrix
0801080
100μg
Each for $296.50
Add to cart
 
22-157-359
View Documents
ZeptoMetrix
0801136
1mg
Each for $2,030.00
Add to cart
 
Description

Description

Host Species: Mouse
Clone: 39/5.4A
Isotype: IgG1
Species Reactivity: Hu

Enzyme Immunoassay, Immunofluorescence, Western Blotting

Specifications

Specifications

HIV-I p24
Purified
RUO
Monoclonal
39/5.4A
Mouse
IgG1
Primary
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit