Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV1 gp120 Goat anti-Virus, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA17218

Catalog No. PA17218


Add to cart

Description

Description

PA1-7218 detects the HIV1 gp120. This antibody does not cross-react with human T or B cells. PA1-7218 has been successfully used in Western blot and ELISA procedures. The PA1-7218 immunogen is purified native gp120 from strain IIIB. Recommended storage of up to 6 months at 2-8°C. For long term storage, aliquot and freeze. avoiding freeze/thaw cycles.

Human immunodeficiency virus type 1 (HIV-1) entry into target cells is directed by the envelope (Env) glycoproteins, which are present on the surface of HIV-1 virion or infected cells in the form of trimers consisting of gp120/gp41 complexes. The surface subunit, gp120, initiates the entry process by interacting sequentially with the CD4 receptor and a co-receptor CCR5 or CXCR4, thereby inducing a conformational change that allows the transmembrane (TM) gp41 subunit to mediate fusion between viral and target cell membranes. Cleavage of Env into its gp120 and gp41 components is necessary for activation of its fusogenic activity.
Specifications

Specifications

HIV1 gp120
Polyclonal
Unconjugated
Liquid
Goat
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Virus
ELISA, Western Blot
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Human immunodeficiency virus; HIV Glycoprotein 120; HIV-1 Glycoprotein 120
Purified native gp120 from strain IIIB.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit