Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV1 p24 Goat anti-Virus, Biotin, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA173095

Catalog No. PA173095


Add to cart

Description

Description

The PA1-73095 antibody reacts with HIV-1 p24. PA1-73095 has been successfully used in Western blot applications. The PA1-73095 antibody was raised against purified native p24 from strain IIIB. Recommended short term storage of less than six months, keep at 2-8°C. For long term storage, store at -20°C.

HIV-1 Gag p24 is a capsid protein that constitutes the core of the AIDS virus HIV-1 and is produced by the digestion of its precursor Gag p55 by HIV-1 protease. The HIV-1 Gag p24 protein is indispensable to the reproduction of AIDS virus, and constitutes an essential element for the AIDS virus particle construction. HIV-1 Gag p24 is detectable from the early stage of AIDS virus infection, and is used as a marker for observation of the development in the patient’s condition after treatment, as it indicated the amount of virus in the blood.
Specifications

Specifications

HIV1 p24
Polyclonal
Biotin
Liquid
Goat
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Virus
Western Blot
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Human immunodeficiency virus; HIV1gp1; HIV p24
Purified native p24 from strain IIIB.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit