Learn More
HIV1 Reverse Transcriptase Mouse anti-Virus, Biotin, Clone: AS35, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112313
Catalog No. 01-674-284
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
HIV-1 reverse transcriptase is an RNA-dependent DNA polymerase of HIV-1(AIDS virus), subtype B origin. It also has RNaseH activity and is an enzyme indispensable to the reproduction of AIDS virus.
Specifications
Specifications
HIV1 Reverse Transcriptase
Monoclonal
1 mg/mL
Dulbecco′s PBS with 0.08% sodium azide
Human immunodeficiency virus; HIV RT
Recombit HIV RT protein.
Protein A/G
RUO
Antibody
Monoclonal
ELISA, Immunoprecipitation
AS35
Biotin
Liquid
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Virus