Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIV1 Virions Polyclonal Antibody, Biotin, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA173092
Description
The PA1-73092 antibody reacts with HIV-1 Virions. Does not react wtih T4+ human lymphocytes. PA1-73092 has been successfully used in Immunofluorescence, immunohistochemistry, ELISA, and western blot applications. The PA1-73092 antibody was raised against HIV-1 isolate. Store at 4oC short term. For long term storage, store at -20oC, avoiding freeze/thaw cycles.
HIV1 Virions Polyclonal antibody specifically detects HIV1 Virions in Virus samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
HIV1 Virions | |
Polyclonal | |
Biotin | |
HIV-1; HIV-1 Virions | |
Human confirmed HIV-1 isolate. | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Goat | |
purified | |
RUO | |
Virus | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction