Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV2 gp36 Mouse anti-Virus, Clone: 3B10/G2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA17589

Catalog No. 01-670-323


Add to cart

Description

Description

MA1-7589 detects HIV from virus samples. MA1-7589 has been successfully used in ELISA, Immunoprecipitation and Western blot procedures. The MA1-7589 immunogen is a purified recombit HIV2 gp36 protein. Store at 4°C short term. For long term storage, aliquot and freeze at -80°C.

Specifications

Specifications

HIV2 gp36
Monoclonal
1 mg/mL
PBS with 0.01% sodium azide
Human immunodeficiency virus; HIV-2; HIV-2 gp36
Purified recombit HIV2 gp36 protein.
Protein G
RUO
Antibody
Monoclonal
ELISA, Western Blot
3B10/G2
Unconjugated
Liquid
Mouse
IgG2a
100 μg
4°C or -80°C if preferred
Primary
Virus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit