Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-A2 Mouse anti-Human, BB515, Clone: BB7.2, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564577

Catalog No. BDB564577


Add to cart

Description

Description

The monoclonal antibody BB7.2 specifically binds to the α subunit of the human leukocyte antigen-A2 (HLA-A2), a class I molecule of the major histocompatibility complex (MHC). The MHC gene locus encodes a group of highly polymorphic, cell-surface proteins that play a broad role in the immune response to protein antigens. MHC molecules function by binding and presenting small antigenic protein fragments to antigen-specific receptors expressed by T cells (TCR). Human ( human leukocyte antigen /HLA) MHC molecules comprise two major classes, MHC class I and class II. Functionally, class I MHC molecules can bind peptides derived from intracellular antigens (eg, viral and some bacterial antigens) that are specifically recognized by CD8+ T cells, and class II MHC molecules recognize antigens derived from pathogens multiplying in intracellular vesicles, and those derived from ingested extracellular bacteria. When presented on the cell surface by the MHC class II molecules, these antigens are recognized by CD4+ T cells. TCR recognize both processed peptides bound to MHC, as well as regions of the MHC molecule itself. CD4 and CD8 accessory molecules strengthen formation of the TCR-MHC complex through their interaction with non-polymorphic regions of the MHC molecule.

Specifications

Specifications

HLA-A2
BB7.2
BB515
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human
Flow Cytometry
0.2mg/mL
HLA class I histocompatibility antigen A2 alpha chain
Affinity Purified
IgG2b, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit