Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

major histocompatibility complex, class II, DO alpha (B02), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human HLA-DOA protein.

Manufacturer:  Abnova Corporation H00003111B02

Catalog No. 89-005-533


Add to cart

Description

Description

HLA-DOA belongs to the HLA class II alpha chain paralogues. HLA-DOA forms a heterodimer with HLA-DOB. The heterodimer, HLA-DO, is found in lysosomes in B cells and regulates HLA-DM-mediated peptide loading on MHC class II molecules. In comparison with classical HLA class II molecules, this gene exhibits very little sequence variation, especially at the protein level. [provided by RefSeq

Sequence: MALRAGLVLGFHTLMTLLSPQEAGATKADHMGSYGPAFYQSYGASGQFTHEFDEEQLFSVDLKKSEAVWRLPEFGDFARFDPQGGLAGIAAIKAHLDILVERSNRSRAINVPPRVTVLPKSRVELGQPNILICIVDNIFPPVINITWLRNGQTVTEGVAQTSFYSQPDHLFRKFHYLPFVPSAEDVYDCQVEHWGLDAPLLRHWELQVPIPPPDAMETLVCALGLAIGLVGFLVGTVLIIMGTYVSSVPR
Specifications

Specifications

major histocompatibility complex, class II, DO alpha
Unconjugated
No additive
NM_002119
HLA-DNA|HLA-DZA|HLADZ
Mouse
50 uL
Membrane Receptors
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human HLA-DOA protein.
HLA-DOA
Serum
HLA-DOA
HLA-DOA (NP_002110, 1 a.a. ~ 250 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
3111
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit