Learn More
anti-HLA-DR, Alex Fluor 405, Clone: L203, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB4869T025
 View more versions of this product
Catalog No. FAB4869T025
Add to cart
Specifications
Specifications
HLA-DR
L203
Flow Cytometry 5 uL/10^6 cells
P01903
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLADR, HLA-DRA, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MHC Class II DR, MLRW
Mouse
IgG1
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human HLA-DR.
Flow Cytometry
Alexa Fluor 594
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Supernatant
HLA-DRA
RPMI 8866 human lymphoblastoid cells, Accession # P01903
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3122
Human