Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-HLA-DR, Alex Fluor 405, Clone: L203, R&D Systems
Mouse Monoclonal Antibody
Manufacturer: R&D Systems FAB4869T100
Specifications
HLA-DR | |
L203 | |
Flow Cytometry 5 uL/10^6 cells | |
P01903 | |
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLADR, HLA-DRA, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MHC Class II DR, MLRW | |
Mouse | |
IgG1 | |
100 Tests | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
Monoclonal | |
Detects human HLA-DR. |
Flow Cytometry | |
Alexa Fluor 594 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Supernatant | |
HLA-DRA | |
RPMI 8866 human lymphoblastoid cells Accession # P01903 | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
3122 | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only