Learn More
anti-HLA-DR, Alex Fluor 647, Clone: L203, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB4869R025
 View more versions of this product
Catalog No. FAB4869R025
Add to cart
This item is not returnable. View return policy
Description
Description
HLA-DR Monoclonal antibody specifically detects HLA-DR in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
HLA-DR
L203
Flow Cytometry 5 uL/10^6 cells
P01903
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLADR, HLA-DRA, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MHC Class II DR, MLRW
Mouse
IgG1
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human HLA-DR.
Flow Cytometry
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Supernatant
HLA-DRA
RPMI 8866 human lymphoblastoid cells Accession # P01903
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3122
Human