Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HLA-DR, Alex Fluor 647, Clone: L203, R&D Systems™

Mouse Monoclonal Antibody

$177.50 - $345.50

Specifications

Antigen HLA-DR
Immunogen RPMI 8866 human lymphoblastoid cells Accession # P01903
Clone L203
Applications Flow Cytometry
Conjugate Alexa Fluor 647
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB4869R025
View Documents
R&D Systems
FAB4869R025
25 Tests
Each for $177.50
Add to cart
 
FAB4869R100
View Documents
R&D Systems
FAB4869R100
100 Tests
Each for $345.50
Add to cart
 
Description

Description

HLA-DR Monoclonal antibody specifically detects HLA-DR in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

HLA-DR
L203
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Mouse
IgG1
3122
RUO
RPMI 8866 human lymphoblastoid cells Accession # P01903
Flow Cytometry
Supernatant
P01903
Monoclonal
Primary
Detects human HLA-DR.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit