Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-HLA-DR, BV786; Clone: G46-6; BD
Supplier: BD Biosciences 564041
Description
Applications:
Flow cytometry
Specifications
HLA-DR | |
Monoclonal | |
Brilliant Violet 786 | |
Affinity Purified | |
Primary | |
Store undiluted at 4deg.C and protected from prolonged exposure to light. Do not freeze |
Flow Cytometry | |
G46-6 | |
Mouse | |
RUO | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction