Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HLA-DR Alexa Fluor 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4869V025
This item is not returnable.
View return policy
Description
HLA-DR Monoclonal specifically detects HLA-DR in Human samples. It is validated for Flow Cytometry.Specifications
HLA-DR | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLADR, HLA-DRA, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MHC Class II DR, MLRW | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3122 | |
Human | |
Supernatant |
Flow Cytometry | |
L203 | |
Flow Cytometry 5 uL/10^6 cells | |
P01903 | |
HLA-DRA | |
RPMI 8866 human lymphoblastoid cells Accession # P01903 | |
25 Tests | |
Primary | |
Detects human HLA-DR. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction