Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-HLA-DR, PerCP, Clone: L203, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems FAB4869C100
This item is not returnable.
View return policy
Description
HLA-DR Monoclonal specifically detects HLA-DR in Human samples. It is validated for Flow Cytometry.Specifications
HLA-DR | |
Monoclonal | |
PerCP | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLADR, HLA-DRA, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MHC Class II DR, MLRW | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3122 | |
Human | |
Purified |
Flow Cytometry | |
L203 | |
Flow Cytometry 10 uL/10^6 cells | |
P01903 | |
HLA-DRA | |
RPMI 8866 human lymphoblastoid cells Accession # P01903 | |
100 Tests | |
Primary | |
Detects human HLA-DR. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title