Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-DR Mouse anti-Human, V450, Clone: L243 (G46-6), BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 642276
Description
Anti–HLA-DR, clone L243, is derived from the hybridization of mouse NS-1/1-Ag4 myeloma cells with spleen cells from BALB/c mice immunized with the human lymphoblastoid B-cell line RPMI 8866.
Anti–HLA-DR recognizes a human class II major histocompatibility complex (MHC) antigen. The antigen is a transmembrane glycoprotein composed of α- and β-subunits that have molecular weights of 36 and 27 kilodaltons (kd), respectively.
Specifications
HLA-DR | |
Monoclonal | |
50μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
100 Tests | |
Immunology | |
Human | |
IgG2a κ |
Flow Cytometry | |
L243 (G46-6) | |
Violet Fluor 450 (V450) | |
Mouse | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction