Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-G, Mouse, Clone: 2A12, Abnova™

Mouse monoclonal antibody raised against synthetic peptide of HLA-G.

Manufacturer:  Abnova Corporation MAB0966

Catalog No. 89-024-821


Add to cart

Description

Description

HLA-G belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA-G is expressed on fetal derived placental cells. The heavy chain is approximately 45kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domain, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exon 6 encodes the cytoplasmic tail. [provided by RefSeq

Specifications

Specifications

HLA-G
2A12
Unconjugated
The optimal working dilution should be determined by the end user.
HLA-G
MHC-G
Murine
IgG1
RUO
Primary
3135
Human
Flow Cytometry, Immunohistochemistry, Western Blot
1 mg/mL
Mouse monoclonal antibody raised against synthetic peptide of HLA-G.
In PBS, pH 7.4 (0.09% sodium azide)
Liquid
HLA-G
A synthetic peptide (conjugated with OVA) corresponding to C-terminus of human soluble HLA-G5 and HLA-G6.
100 ug
Store at 4°C
Monoclonal
This antibody allows to discriminate between soluble HLA-G protein yielded by shedding from membrane-bound HLA-G forms that do not contain the intron 4-encoded epitope and soluble HLA-G5/HLA-G6 produced from intron 4-retaining alternatively spliced mRNAs.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit