Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLX Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124359

Catalog No. PIPA124359


Add to cart

Description

Description

PA1-24359 detects HLX1 in human samples. PA1-24359 has been successfully used in Western blot procedures. The PA1-24359 immunogen is a synthetic peptide region of human HLX1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

H2.0-like homeo box 1; H2.0 (Drosophilia)-like homeo box-1.
Specifications

Specifications

HLX
Polyclonal
Unconjugated
HLX
Liquid
HLX
Synthetic peptide corresponding to a region of human HLX1.
Protein A/G
RUO
Antibody
Polyclonal
Human
Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q14774
HB24, HLX1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3142
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit