Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-hmg-3, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals 48690002
Description
hmg-3 Polyclonal antibody specifically detects hmg-3 in C. elegans samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
hmg-3 | |
1 mg/mL | |
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Affinity Purified | |
Rabbit | |
IgG | |
0.05 ml | |
modENCODE Antibodies | |
Primary | |
172250 | |
C. elegans |
ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative | |
Protein HMG-3 | |
In vivo generated recominant protein fragment | |
Immunogen affinity purified | |
RUO | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Polyclonal | |
Caenorhabditis elegans. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only