Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGA1 Goat anti-Human, Mouse, Rat, Porcine, Canine, Hamster, Monkey, Rabbit, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP150394

Catalog No. NBP150394

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

HMGA1 Polyclonal antibody specifically detects HMGA1 in Human, Mouse, Rat, Porcine, Canine, Hamster, Monkey, Rabbit samples. It is validated for ELISA, Immunohistochemistry (Paraffin).
Specifications

Specifications

HMGA1
0.5 mg/mL
ELISA 1:32000, Immunohistochemistry-Paraffin 4 ug/ml
Affinity Purified
HMGA1
Synthetic peptide C-LASKQEKDGTEK from an internal region of human HMGA1 (NP_665906.1; NP_665908.1; NP_002122.1; NP_665909.1; NP_665912.1; NP_665910.1).
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Canine, Hamster, Human, Monkey, Mouse, Porcine, Rabbit, Rat
ELISA, Immunohistochemistry (Paraffin)
Unconjugated
TBS (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
high mobility group AT-hook 1, High mobility group AT-hook protein 1, High mobility group protein A1, high mobility group protein HMG-I/HMG-Y, High mobility group protein R, high-mobility group (nonhistone chromosomal) protein isoforms I and Y, HMGA1A, HMG-I(Y), HMGIYMGC12816, HMG-R, MGC4242, MGC4854, nonhistone chromosomal high-mobility group protein HMG-I/HMG-Y
Goat
IgG
0.05 mg
Cell Cycle and Replication
Primary
3159
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only